missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, Reagent Grade

Product Code. 11326748
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326748 1 kg 1kg
11316748 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326748 Supplier Thermo Scientific Alfa Aesar Supplier No. 013417.A1
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6108-17-4 | C2H7LiO4 | 102.014 g/mol

Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.014
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 54°C to 57°C
Color White
Physical Form Crystals
Quantity 1 kg
Density 1.300 g/mL
Beilstein 3564320
Merck Index 14,5521
Solubility Information Freely soluble in water and alcoholSoluble in water and ethanol.
Formula Weight 102.01 (65.98 Anhydrous)
Grade Reagent
Chemical Name or Material Lithium acetate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.