missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum di-n-butylamide, 0.16M solution in 1,2-Dimethoxyethane, Thermo Scientific Chemicals

Product Code. 13515040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13515040 500 mL Glass bottle 500mL
13505040 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13515040 Supplier Thermo Scientific Acros Supplier No. 449195000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15405-86-4
Molecular Formula C32H72AlLiN4
Molecular Weight (g/mol) 546.87
InChI Key ISBVOXPIPNIBOM-UHFFFAOYSA-N
Synonym lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
PubChem CID 57369578
IUPAC Name lithium;tetrakis(dibutylamino)alumanuide
SMILES [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC

Specifications

Melting Point -54°C
Quantity 500 mL
Flash Point -6°C
Formula Weight 546.87
Packaging Glass bottle
Specific Gravity 0.86
Density 0.8600g/mL
Chemical Name or Material Lithium Aluminum di-n-butylamide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium aluminium di-n-butylamide, 0.16M solution in 1,2-dimethoxyethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.