missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 95%, tablets (ca. 0.6 g)

Product Code. 10063610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Tamaño de la unidad:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
10063610 25 g 25g
10349310 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.
Código de producto. 10063610 Proveedor Thermo Scientific Acros N.º de proveedor 304530250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto es un intermedio aislado transportado de acuerdo con el Reglamento (CE) 1907/2006 (REACH). Todos los clientes deben completar una declaración antes de la entrega.

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 16853-85-3
Molecular Formula AlH4Li
Molecular Weight (g/mol) 37.95
MDL Number MFCD00011075
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445
IUPAC Name lithium(1+) alumanuide
SMILES [Li+].[AlH4-]

Especificaciones

Melting Point 125.0°C
Color White to Gray
Physical Form Pellets
Quantity 25 g
Assay Percent Range 94% min. (LiAlH4)
Linear Formula LiAlH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Solubility Information Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd
Formula Weight 37.95
Percent Purity 95%
Density 0.9
Chemical Name or Material Lithium Aluminum hydride, 95%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Lithium aluminium hydride
Palabra de advertencia
  • Danger
Clasificación
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Consejos de prudencia
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.