missing translation for 'onlineSavingsMsg'
Learn More

Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane

Product Code. 11328075
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328075 100 mL 100mL
11338075 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328075 Supplier Thermo Scientific Alfa Aesar Supplier No. L08269.AE
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15405-86-4 | C32H72AlLiN4 | 546.878 g/mol

Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.

Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15405-86-4
Molecular Formula C32H72AlLiN4
Molecular Weight (g/mol) 546.878
MDL Number MFCD00051918
InChI Key ISBVOXPIPNIBOM-UHFFFAOYSA-N
Synonym lithium aluminum di-n-butylamide, lithium tetrakis dibutylamino alumanuide, lithium aluminum di-n-butylamide, 0.16m soln. in 1,2-dimethoxyethane
PubChem CID 57369578
IUPAC Name lithium;tetrakis(dibutylamino)alumanuide
SMILES [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC

Specifications

Melting Point -54°C
Density 0.86
Linear Formula LiAl[N(CH2CH2CH2CH3)2]4
Quantity 100 mL
UN Number UN1993
Sensitivity Air and moisture sensitive
Formula Weight 546.87
Chemical Name or Material Lithium aluminum di-n-butylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.