Learn More
Lithium aluminum hydride, 95% min, Thermo Scientific™
Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen
140.00 €
Chemical Identifiers
| CAS | 16853-85-3 |
|---|---|
| Molecular Formula | AlH4Li |
| Molecular Weight (g/mol) | 37.95 |
| MDL Number | MFCD00011075 |
| InChI Key | OCZDCIYGECBNKL-UHFFFAOYSA-N |
| Synonym | lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride |
| PubChem CID | 21226445 |
| IUPAC Name | lithium(1+) alumanuide |
| SMILES | [Li+].[AlH4-] |
Description
Reducing agent for pharmaceuticals, perfumes and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizationsLithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It is also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 16853-85-3 | |
| 37.95 | |
| OCZDCIYGECBNKL-UHFFFAOYSA-N | |
| 21226445 | |
| [Li+].[AlH4-] |
| AlH4Li | |
| MFCD00011075 | |
| lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
| lithium(1+) alumanuide |
Specifications
| 16853-85-3 | |
| MFCD00011075 | |
| 14,349 | |
| Decomposes in water; Soluble in ether,diethyl ether and terahydrofuran. | |
| [Li+].[AlH4-] | |
| 37.95 | |
| 37.95 | |
| Odorless | |
| 0.917 |
| AlH4Li | |
| UN1410 | |
| lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride | |
| OCZDCIYGECBNKL-UHFFFAOYSA-N | |
| lithium(1+) alumanuide | |
| 21226445 | |
| ≥95% | |
| Air and moisture sensitive | |
| Lithium aluminum hydride |
Safety and Handling
GHS H Statement
H260-H301-H314-H318;
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H500
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM ALUMINUM HYDRIDE
missing translation for 'einecsNumber' : 240-877-9
missing translation for 'rtecsNumber' : BD0100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.