missing translation for 'onlineSavingsMsg'
Learn More

Lithium amide, 95%, Thermo Scientific™

Product Code. 10471241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
Product Code. Quantity Packaging unitSize
10471241 100 g Glass bottle 100g
10471051 5 g Glass bottle 5g
10296730 500 g Glass bottle 500g
3 options
This item is not returnable. View return policy

Product Code. 10471241

Brand: Thermo Scientific Acros 199861000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-89-0
MDL Number MFCD00011093
InChI Key AFRJJFRNGGLMDW-UHFFFAOYSA-N
Synonym lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
PubChem CID 24532
IUPAC Name lithium;azanide
SMILES [Li+].[NH2-]

Specifications

Boiling Point 430°C
Melting Point 373°C
Color White through Gray
Physical Form Fine Powder
Quantity 100 g
Assay Percent Range 94% min. (LiNH2)
Molecular Formula H2LiN
Linear Formula LiNH2
Fieser 01,600; 03,178; 05,392; 13,159; 17,311
Merck Index 15, 5578
Solubility Information Solubility in water: vigorous reaction. Other solubilities: forming lioh and nh3,insoluble in anhydr. ether,benzene and toluene
Molecular Weight (g/mol) 22.95
Formula Weight 22.95
Percent Purity 95%
Packaging Glass bottle
Specific Gravity 1.17
Density 1.1700g/mL
Chemical Name or Material Lithium amide, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.