missing translation for 'onlineSavingsMsg'
Learn More

Lithium arsenate, 99%

Product Code. p-7050727
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11308118

Brand: Thermo Scientific Alfa Aesar 039350.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 13478-14-3 | AsLi3O4 | 159.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-14-3
Molecular Formula AsLi3O4
Molecular Weight (g/mol) 159.738
MDL Number MFCD00053998
InChI Key VKJKOXNPYVUXNC-UHFFFAOYSA-K
Synonym lithium arsenate, trilithium arsenate, trilithium arsorate, arsenic acid h3aso4 , trilithium salt, acmc-20akn0, aso4.3li, arsenic acid h3aso4 , lithium salt 1:3, trilithium 1+ ion arsenate, arsenic acid h3aso4 ,lithium salt 1:3
PubChem CID 166828
IUPAC Name trilithium;trioxido(oxo)-$l^{5}-arsane
SMILES [Li+].[Li+].[Li+].[O-][As](=O)([O-])[O-]

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN1557
Solubility Information Soluble in dilute acetic acid; Insoluble in water
Formula Weight 159.74
Percent Purity 99%
Density 3.07 g/mL
Chemical Name or Material Lithium arsenate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.