missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trifluoromethylsulfonyl)imide, 98+%

Product Code. 11382379
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382379 50 g 50g
11372379 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382379 Supplier Thermo Scientific Alfa Aesar Supplier No. H27307.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90076-65-6 | C2F6LiNO4S2 | 287.075 g/mol

Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium bis(trifluoromethylsulfonyl)imide is used in the preparation of chiral imidazolium salt through an anion metathesis of the corresponding triflate organic electrolyte-based lithium batteries. It finds application in the preparation of rare-earth Lewis acid catalysts. It is also useful in primary and secondary lithium cells using organic liquid electrolytes and polymer batteries.

Solubility
Soluble in water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90076-65-6
Molecular Formula C2F6LiNO4S2
Molecular Weight (g/mol) 287.075
MDL Number MFCD00210017
InChI Key QSZMZKBZAYQGRS-UHFFFAOYSA-N
Synonym lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimide
PubChem CID 3816071
IUPAC Name lithium;bis(trifluoromethylsulfonyl)azanide
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 234°C to 238°C
Linear Formula (CF3SO2)2NLi
Quantity 50 g
UN Number UN2923
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 287.08
Percent Purity ≥98%
Chemical Name or Material Lithium bis(trifluoromethylsulfonyl)imide
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Lithium bis(trifluoromethylsulfonyl)imide
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Gefahrenhinweis
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Lithium bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.