missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, Thermo Scientific™

Product Code. 15761809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15761809 100 mL AcroSeal™ Glass bottle 100mL
10472293 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15761809 Supplier Thermo Scientific Acros Supplier No. 380231000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4039-32-1 , 108-87-2
MDL Number MFCD00008261
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
PubChem CID 2733832
IUPAC Name lithium;bis(trimethylsilyl)azanide
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C

Spécification

Color Colorless to Yellow
Density 0.7700g/mL
Flash Point −3°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C6H18LiNSi2
Quantity 100 mL
Specific Gravity 0.77
Solubility Information Solubility in water: hydrolysis
Molecular Weight (g/mol) 167.33
Formula Weight 167.33
Physical Form Liquid
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.