missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trimethylsilyl)amide, 1.0M sol. in methyl tert-butyl ether, AcroSeal™

Product Code. 10183391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10183391 800 mL AcroSeal™ Glass Bottle 800mL
10052011 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10183391 Supplier Thermo Scientific Acros Supplier No. 345678000
  • ONLINE EXCLUSIVE PRICE
    775.00 €
    723.00 € / 800mL
    Save 52.00 €
    6% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 4039-32-1 | C6H18LiNSi2 | 167.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
Name Note 1.0M Solution in Methyl tert-Butyl Ether
CAS 4039-32-1, 1634-04-4
Color Brown to Yellow
Density 0.8000g/mL
Boiling Point 55.0°C to 56.0°C
Flash Point −28°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H18LiNSi2
MDL Number MFCD00008261
Quantity 800 mL
Fieser 04,296; 05,393; 07,197; 12,280; 13,165; 14,194
Specific Gravity 0.8
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C
IUPAC Name lithium;bis(trimethylsilyl)azanide
Molecular Weight (g/mol) 167.33
PubChem CID 2733832
Formula Weight 167.33
Percent Purity 21 to 25% active base (as LiNSi)
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.