missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trimethylsilyl)amide, 15% in 2-MeTHF/Ethylbenzene, AcroSeal™, Thermo Scientific™

Product Code. 12327173 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
800mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12327173 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12327173 Supplier Thermo Scientific Acros Supplier No. 446118000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4039-32-1, 96-47-9, 100-41-4
Quantity 800 mL
Assay Percent Range 15%
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
Flash Point −11°C
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
Solubility Information Solubility in water: decomposes
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C
IUPAC Name lithium;bis(trimethylsilyl)azanide
Molecular Weight (g/mol) 167.33
PubChem CID 2733832
Formula Weight 167.33
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.856
Density 0.8560g/mL
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.