missing translation for 'onlineSavingsMsg'
Learn More

Lithium Borohydride, TRC

Product Code. 30483771
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483771 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483771 Supplier Toronto Research Chemicals Supplier No. TRCL4690802.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Borohydride
CAS 16949-15-8
Quantity 2.5 g
Molecular Formula BH4Li
Synonym Lithium Tetrahydroborate (1:1), Lithium Boron Hydride, Lithium Hydroborate (LiBH4), Lithium Tetrahydroborate
InChI Formula InChI=1S/BH4.Li/h1H4;/q-1;+1
SMILES [Li+].[BH4-]
IUPAC Name lithium;boranuide
Molecular Weight (g/mol) 21.78
Formula Weight 22.0566
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.