missing translation for 'onlineSavingsMsg'
Learn More

Lithium bromide, 99+%, for analysis, anhydrous

Product Code. 15234674 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Unit Size:
100g
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15234674

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7550-35-8 | BrLi | 86.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.84
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265.0°C
Melting Point 550.0°C
Color Gray to Black
pH 6 to 12 (4% soln. at 20°C)
Physical Form Powder
Quantity 10 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiBr
Fieser 01,604; 02,245; 04,297
Merck Index 15,5582
Solubility Information Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol
Formula Weight 86.84
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5% max. (400°C)
Specific Gravity 3.46
Density 3.4600g/mL
Chemical Name or Material Lithium bromide, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.