Learn More
Lithium bromide, anhydrous, 99% min
Description
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange mediumLithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1265°C |
| Melting Point | 550°C |
| Color | White |
| Physical Form | Crystals |
| Quantity | 500 g |
| Assay Percent Range | 99% min. |
| Solubility Information | Very soluble in water and alcohol. Soluble in methanol,acetone,and glycol. Slightly soluble in pyridine. |
| Formula Weight | 86.85 |
| Sensitivity | Hygroscopic |
| Density | 3.464 g/cm3 at 20°C |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.