missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide, puriss p.a, ≥99.0% (dried material, AT), Honeywell Fluka™

Product Code. 15606050 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15606050 25 g Glass bottle 25g
15696040 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15606050 Supplier Honeywell-Fluka Supplier No. 6246425G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (dried material, AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Color Colorless
Physical Form Solid
Quantity 25 g
Linear Formula LiBr
UN Number NONH for all modes of transport
Formula Weight 86.85g/mol
Percent Purity ≥99.0% (dried material, AT)
Odor Odorless
Packaging Glass bottle
Density 3.46g/cm3
Chemical Name or Material Lithium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.