missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, 99.999%, (trace metal basis), extra pure

Product Code. 10239833 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10239833

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 720.0°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999%
Linear Formula Li2CO3
Fieser 01,606; 03,183; 04,192; 05,396
Merck Index 15, 5583
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids,practically insoluble in alcohol
IUPAC Name dilithium;carbonate
Formula Weight 73.88
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Density 2.11g/mL
Chemical Name or Material Lithium carbonate, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.