missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, ACS, 99.0% min

Product Code. 11303278
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11303278

Brand: Thermo Scientific Alfa Aesar 036225.36

This item is not returnable. View return policy

Maximum level of impurities: Insoluble in dilute hydrochloric acid 0.01%, Cl 0.005%, NO3 5ppm, Sulfur compounds (as SO4) 0.2%, NH4 5ppm, Heavy Metals (as Pb) 0.002%, Fe 0.002%, Ca 0.01%, K 0.01%, Na 0.1% | CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1310°C (decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water and acid. Insoluble in acetone,ammonia and alcohol.
IUPAC Name dilithium;carbonate
Formula Weight 73.9
Grade ACS
Odor Odorless
Appearance White crystalline powder
Density 2.11 g/cm3 at 20°C
Chemical Name or Material Lithium carbonate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.