missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, ACS, 99.0% min

Product Code. 11313278
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313278 2 kg 2kg
11303278 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313278 Supplier Thermo Scientific Alfa Aesar Supplier No. 036225.A3
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximum level of impurities: Insoluble in dilute hydrochloric acid 0.01%, Cl 0.005%, NO3 5ppm, Sulfur compounds (as SO4) 0.2%, NH4 5ppm, Heavy Metals (as Pb) 0.002%, Fe 0.002%, Ca 0.01%, K 0.01%, Na 0.1% | CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1310°C (decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 2 kg
Odor Odorless
Appearance White crystalline powder
Assay Percent Range 99.0% min.
Density 2.11
Solubility Information Soluble in water and acid. Insoluble in acetone,ammonia and alcohol.
IUPAC Name dilithium(1+) carbonate
Formula Weight 73.9
Grade ACS
Chemical Name or Material Lithium carbonate, For analysis ACS
Show More Show Less
compliance-icons
Product Identifier
  • Lithium carbonate, ACS
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Lithium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.