missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Certified AR for Analysis, Fisher Chemical

Product Code. 10695492 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10695492 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10695492 Supplier Fisher Chemical Supplier No. L/2105/48
  • ONLINE EXCLUSIVE PRICE
    181.00 €
    179.00 € / 100g
    Save 2.00 €
    1% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Specifications

Melting Point 720°C
CAS 554-13-2
Color White
pH 10 to 11
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula CLi2O3
MDL Number MFCD00011084
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES [Li+].[Li+].[O-]C([O-])=O
IUPAC Name dilithium;carbonate
Molecular Weight (g/mol) 73.89
PubChem CID 11125
ChEBI CHEBI:6504
Formula Weight 73.88g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa 610
Show More Show Less
compliance-icons
Product Identifier
  • Lithium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.