missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Solstice

Product Code. 15681230
Change view
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
100 kg
Unit Size:
100g
100kg
1kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15681230 6 x 1 kg Pack of 6
15651230 100 g 100g
15671230 1 kg 1kg
15661230 100 kg 100kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15681230 Supplier Honeywell-Fluka Supplier No. 130106X1KG
 Restricted Item
Estimated Shipment: 18-06-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Quantity 6 x 1 kg
Packaging Plastic Bottle
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Formula Weight 73.89
Percent Purity ≥99%
Grade purum
Chemical Name or Material Lithium Carbonate
compliance-icons
Product Identifier
  • Lithium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lithium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.