missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, puriss p.a, ≥99.0% (T), For Microscopy, Solstice

Product Code. 15626050
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15626050 500 g Poly bottle 500g
15616050 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15626050 Supplier Honeywell-Fluka Supplier No. 62470500G
  • ONLINE EXCLUSIVE PRICE
    178.00 €
    160.93 € / 500g
    Save 17.07 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, reagent (for microscopy),≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1265°C (1013hPa)
Melting Point 550°C
Color Colorless
pH 9 to 11
Physical Form Solid
Quantity 500 g
Identification Passes Test
Odor Odorless
Packaging Poly bottle
Product Line Solstice
Assay Percent Range ≥99.0%
Density 3.46g/cm3
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
InChI Formula 1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
Formula Weight 73.89g/mol
Percent Purity ≥99.0% (T)
Concentration or Composition (by Analyte or Components) Chloride: 20 mg/kg, Nitrate: 5 mg/kg, Sulfate: 50 mg/kg, Ca: 50 mg/kg, Cd: 5 mg/kg, Co: 5 mg/kg, Cr: 5 mg/kg, Cu: 5 mg/kg, Fe: 5 mg/kg, K: 50 mg/kg, Mg: 10 mg/kg, Mn: 5 mg/kg, NH4+: 5 mg/kg, Na: 50 mg/kg, Ni: 5 mg/kg, Pb: 5 mg/kg, Zn: 5 mg/kg
Manufacturing Origin Germany
Assay Titration
Chemical Name or Material Lithium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lithium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.