missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, TraceSELECT™, ≥99.998% (metals basis)

Product Code. 15686040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Poly Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15686040 25 g Poly Bottle 25g
15676040 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15686040 Supplier Honeywell-Fluka Supplier No. 6246225G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Color White
pH 9.0 to 10.00 (at 1g/L, 20°C)
Physical Form Crystalline
Quantity 25 g
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
IUPAC Name dilithium;carbonate
Formula Weight 73.89g/mol
Percent Purity ≥99.998% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Density 2.11g/cm3
Chemical Name or Material Lithium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.