missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, TRC

Product Code. 30009725
Change view
Click to view available options
Quantity:
50 mg
5 g
Unit Size:
50mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009725 50 mg 50mg
30006295 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009725 Supplier Toronto Research Chemicals Supplier No. TRCL46910050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula C O3 . 2 Li
Molecular Weight (g/mol) 73.89
Synonym Lithium carbonate, Carbonic Acid Lithium Salt, Carbonic Acid Dilithium Salt, CP 15467-61, Camcolit, Candamide, Carbolith, Carbolithium, Carbonic Acid Lithium Salt, Ceglution, Dilithium Carbonate, Eskalith, Hypnorex, Limas, Liskonum, Lithane, Lithicarb, Lithobid, Lithocarb, Lithonate, Lithotabs, Micalith, NSC 16895, Phasal, Plenur, Priadel, Quilonorm-retard, Quilonum Retard, Teralithe;
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
Formula Weight 74.0168
Chemical Name or Material Lithium Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.