missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride monohydrate, 99.95% (metals basis)

Product Code. 11338017
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338017 25 g 25g
11328017 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338017 Supplier Thermo Scientific Alfa Aesar Supplier No. 022151.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16712-20-2 | ClH2LiO | 60.405 g/mol

Lithium chloride monohydrate is used as a precursor in the production of lithium metal and other lithium compounds. It acts as a welding agent for aluminum, salt bath for heat -treatment by low temperature and soldering techniques. It is also used as a tracer for chemical products viz. denaturation of wine. For absorbers, it is an absorption and disinfection reagent as well as desiccant for air conditioner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16712-20-2
Molecular Formula ClH2LiO
Molecular Weight (g/mol) 60.405
MDL Number MFCD00011078
InChI Key VXJIMUZIBHBWBV-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
IUPAC Name lithium;chloride;hydrate
SMILES [Li+].O.[Cl-]

Specifications

Melting Point >98°C-H2O
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 60.41 (42.39 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.78
Chemical Name or Material Lithium chloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium chloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Lithium chloride, monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.