missing translation for 'onlineSavingsMsg'
Learn More

Lithium citrate tetrahydrate, MedChemExpress

Product Code. 16408726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408726 Supplier Medchem Express Supplier No. HYB1295
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lithium citrate (Litarex) tetrahydrate is the major active ingredient of Garcinia cambogia. Lithium citrate tetrahydrate competitively inhibits ATP citrate lyase with weight loss benefits. Lithium citrate tetrahydrate effective inhibits stones formation and also inhibits HIF, and has antioxidation, anti-inflammation and anti-tumor effects.

Lithium citrate (Litarex) tetrahydrate is the major active ingredient of Garcinia cambogia. Lithium citrate tetrahydrate competitively inhibits ATP citrate lyase with weight loss benefits. Lithium citrate tetrahydrate effective inhibits stones formation and also inhibits HIF, and has antioxidation, anti-inflammation and anti-tumor effects.

Specifications

Chemical Name or Material Lithium citrate tetrahydrate
CAS 6080-58-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H13Li3O11
Quantity 100 mg
Synonym Litarex tetrahydrate
Solubility Information H2O : ≥ 100 mg/mL (354.64 mM)
SMILES O=C(CC(C(O[Li])=O)(O)CC(O[Li])=O)O[Li].O.O.O.O
Molecular Weight (g/mol) 281.98
Formula Weight 281.98
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.