missing translation for 'onlineSavingsMsg'
Learn More

Lithium dimethylamide, 96%

Product Code. 11399638
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399638 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11399638 Supplier Thermo Scientific Alfa Aesar Supplier No. 044145.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3585-33-9 | C2H6LiN | 51.017 g/mol

It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3585-33-9
Molecular Formula C2H6LiN
Molecular Weight (g/mol) 51.017
MDL Number MFCD00008295
InChI Key YDGSUPBDGKOGQT-UHFFFAOYSA-N
Synonym lithium dimethylamide, lithium dimethylazanide, linme2, acmc-20akms, dimethylamino lithium, lithiodimethylamine, lithium dimethylamide preparation, lithium 1+ dimethylazanide, lithium dimethylamide w/v in hexane
PubChem CID 5152226
IUPAC Name lithium;dimethylazanide
SMILES [Li+].C[N-]C

Specifications

Melting Point (decomposition)
Color White
Odor Amine-like
Linear Formula LiN(CH3)2
Quantity 5 g
UN Number UN3393
Beilstein 3618233
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently with water.
Formula Weight 51.02
Percent Purity 96%
Physical Form Powder or Lumps
Chemical Name or Material Lithium dimethylamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.