missing translation for 'onlineSavingsMsg'
Learn More

Lithium Dodecyl Sulfate, Ultra Pure, 5g, MP Biomedicals

Product Code. 17861391 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17861391 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17861391 Supplier MP Biomedicals Supplier No. 04800752
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2044-56-6 (LDS Lauryl sulfate lithium salt) Purity: >>>99% Exhibits greater solubility than SDS at lower temperatures, while maintaining similar detergency and wetting ability

Specifications

CAS 2044-56-6
Quantity 5 g
Molecular Formula C12H25LiO4S
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name lithium(1+) dodecyl sulfate
Molecular Weight (g/mol) 272.33
PubChem CID 2735071
Percent Purity 99%
Grade Ultra Pure
Chemical Name or Material Lithium Dodecyl Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.