missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.98%, extra pure

Product Code. 10401051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10401051

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]

Specifications

Boiling Point 1681.0°C
Melting Point 845.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.98%
Linear Formula LiF
Fieser 01,933
Merck Index 15, 5587
Solubility Information Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
IUPAC Name lithium(1+) fluoride
Formula Weight 25.93
Percent Purity 99.98%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.63
Density 2.6300g/mL
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.