missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.98%, extra pure

Product Code. 10626212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10626212 25 g Plastic bottle 25g
10401051 5 g Plastic bottle 5g
10534084 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10626212 Supplier Thermo Scientific Acros Supplier No. 193380250
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]

Specifications

Boiling Point 1681.0°C
Melting Point 845.0°C
Color White
Physical Form Powder
Quantity 25 g
Packaging Plastic bottle
Specific Gravity 2.63
Assay Percent Range 99.98%
Density 2.6300g/mL
Linear Formula LiF
Fieser 01,933
Merck Index 15, 5587
Solubility Information Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
IUPAC Name lithium;fluoride
Formula Weight 25.93
Percent Purity 99.98%
Grade Extra Pure
Chemical Name or Material Lithium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.