missing translation for 'onlineSavingsMsg'
Learn More

Lithium Fluoride, Honeywell Fluka™

Product Code. 15680540 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 15680540

Brand: Honeywell-Fluka 01140100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]

Specifications

Quantity 100 g
Linear Formula LiF
UN Number 3288
Formula Weight 25.94g/mol
Percent Purity ≥99%
Grade purum
Density 2.64g/mL at 25°C
Chemical Name or Material Lithium Fluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.