missing translation for 'onlineSavingsMsg'
Learn More

Lithium granules, 1-6mm (0.04-0.2in), 99%

Product Code. 11326637
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326637 50 g 50g
11336637 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326637 Supplier Thermo Scientific Alfa Aesar Supplier No. 000733.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Sodium is major impurity | CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]

Specifications

Color Gray
Appearance Gray granules
Density 0.53g/cm3 at 20°C
Quantity 50 g
Odor Odorless
Packaging Under Argon
Assay Percent Range 99% (Metals basis)
Form Granular
Chemical Name or Material Lithium
compliance-icons compliance-icons
Product Identifier
  • Lithium granules
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.