missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsDescription
Reduction reagent
Specifications
Specifications
| Melting Point | 680.0°C |
| Color | White to Gray |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 96% min |
| Linear Formula | LiH |
| Fieser | 04,304; 13,165 |
| Merck Index | 15, 5589 |
| Solubility Information | Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether |
| Formula Weight | 7.95 |
| Show More |
Safety and Handling
Product Identifier
- Lithium hydride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?