missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™

Product Code. 10760891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10760891 100 g 100g
10275221 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10760891 Supplier Thermo Scientific Acros Supplier No. 191191000
Quantity Price
1 167.00 € / 100g
10 142.33 € / 100g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7580-67-8
Molecular Formula HLi
Molecular Weight (g/mol) 7.95
MDL Number MFCD00011074
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
PubChem CID 62714
IUPAC Name lithium;hydride
SMILES [H-].[Li+]

Specifications

Melting Point 680.0°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min
Linear Formula LiH
Fieser 04,304; 13,165
Merck Index 15, 5589
Solubility Information Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether
Formula Weight 7.95
Percent Purity 97%
Grade Pure
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Lithium hydride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.