missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydride, 97%, pure, powder (<100 μm)

Product Code. 10526821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10526821 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10526821 Supplier Thermo Scientific Acros Supplier No. 332955000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 7580-67-8 | HLi | 7.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 680.0°C
CAS 7580-67-8
Color White to Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 96% min
Molecular Formula HLi
Linear Formula LiH
MDL Number MFCD00011074
Fieser 04,304; 13,165
Merck Index 15, 5589
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
Solubility Information Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
SMILES [H-].[Li+]
IUPAC Name lithium;hydride
Molecular Weight (g/mol) 7.95
PubChem CID 62714
Formula Weight 7.95
Percent Purity 97%
Grade Pure
Packaging Plastic bag in resealable tin can
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Lithium hydride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.