missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydride, 97+%

Product Code. 15463167
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463167 1000 g 1000g
11422010 50 g 50g
11432010 100 g 100g
11442010 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15463167 Supplier Thermo Scientific Alfa Aesar Supplier No. A17948.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 7580-67-8 | HLi | 7.948 g/mol

Used for preparation of anhydrous Li salts of carboxylic acids prior to reaction with organothium reagents (most often methyl or phenyl) as a useful route to ketones. Also used in the preparation of LiCN by reaction with acetone cyanohydrins. It is also applied as a desiccating and condensing agent and a shielding material in nuclear energy production. It is used in the manufacture of catalysts for polyolefin plastics; in fuels for aircraft, missiles. It is used in metallurgy, as degasifier, deoxidizer, desulfurizer; as grain refiner in non-ferrous metals. Employed as a chemical intermediate for butyllithium polymerization catalyst; in alloys with aluminum or magnesium for aerospace uses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for preparation of anhydrous Li salts of carboxylic acids prior to reaction with organothium reagents (most often methyl or phenyl) as a useful route to ketones. Also used in the preparation of LiCN by reaction with acetone cyanohydrins. It is also applied as a desiccating and condensing agent and a shielding material in nuclear energy production. It is used in the manufacture of catalysts for polyolefin plastics; in fuels for aircraft, missiles. It is used in metallurgy, as degasifier, deoxidizer, desulfurizer; as grain refiner in non-ferrous metals. Employed as a chemical intermediate for butyllithium polymerization catalyst; in alloys with aluminum or magnesium for aerospace uses.

Solubility
Reacts violently with water. Soluble in ether, liquid ammonia. Insoluble in benzene, and toluene.

Notes
Air & Moisture Sensitive. Store away from acids, alcohols, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7580-67-8
Molecular Formula HLi
Molecular Weight (g/mol) 7.948
MDL Number MFCD00011074
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
PubChem CID 62714
IUPAC Name lithium;hydride
SMILES [H-].[Li+]

Specifications

Melting Point 688°C
Quantity 1000 g
Linear Formula LiH
UN Number UN1414
Merck Index 14,5533
Solubility Information Reacts violently with water. Soluble in ether,liquid ammonia. Insoluble in benzene,and toluene.
Formula Weight 7.95
Percent Purity ≥97%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 0.78
Chemical Name or Material Lithium hydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.