missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, anhydrous, 99.995% (metals basis)

Product Code. 11420800
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11420800 5 g 5g
11430800 25 g 25g
11440800 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11420800

Brand: Thermo Scientific Alfa Aesar 044471.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-65-2 | HLiO | 23.95 g/mol

It is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
IUPAC Name lithium(1+) hydroxide
SMILES [Li+].[OH-]

Specifications

Boiling Point 924°C
Melting Point 462°C
Color White
pH 14
Physical Form Powder
Quantity 5 g
UN Number UN2680
Merck Index 14,5534
Solubility Information Soluble in water. Slightly soluble in ethanol and methanol
Formula Weight 23.95
Percent Purity 99.995%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.54 g/mL
Assay (metals basis)
Chemical Name or Material Lithium hydroxide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.