missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium hydroxide monohydrate, 98+%, ACS reagent
Alle ansehen Thermo Scientific Chemicals Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
missing translation for 'unitSize'
100g
2.5kg
500g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Boiling Point | 920.0°C |
| Melting Point | 462.0°C |
| Quantity | 500 g |
| Assay Percent Range | 98+% |
| Linear Formula | LiOH·H2O |
| Merck Index | 14,5534 |
| Solubility Information | Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol |
| IUPAC Name | lithium;hydroxide;hydrate |
| Formula Weight | 41.95 |
| Percent Purity | ≥98% |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Lithium hydroxide monohydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur