missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, battery grade

Product Code. 15417657
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. Afficher la politique du retour.

Code produit. 15417657

missing translation for 'mfr': Thermo Scientific Alfa Aesar H36379.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

Solubility
Soluble in water (216 mg/ml at 20°C), ethanol (freely), and alcohol (slightly).

Notes
Air sensitive and hygroscopic. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Spécification

Quantity 250 g
UN Number UN2680
Solubility Information Soluble in water (216mg/ml at 20°C),ethanol (freely),and alcohol (slightly).
Formula Weight 41.96(23.95 Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Lithium hydroxide monohydrate

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis