missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, TraceSELECT™, ≥99.998% (metals basis), Honeywell™ Fluka™

Product Code. 15641710 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 g
Packaging:
Poly bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 15641710

Brand: Honeywell-Fluka 1951910G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 423°C
CAS 1310-66-3
Color Colorless
pH Alkaline
Physical Form Solid
Quantity 10 g
Molecular Formula H3LiO2
Linear Formula LiOH · H2O
MDL Number MFCD00149772
UN Number UN2680
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES [Li+].O.[OH-]
IUPAC Name lithium(1+) hydrate hydroxide
Molecular Weight (g/mol) 41.96
PubChem CID 168937
Formula Weight 41.96g/mol
Percent Purity ≥99.998% (metals basis)
Grade TraceSELECT
Odor Weak
Packaging Poly bottle
Density ∽1.830g/cu.cm at 20°C
Chemical Name or Material Lithium Hydroxide Monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.