missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide, TRC

Product Code. 30472352
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30472352 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30472352

Brand: Toronto Research Chemicals TRCL46911525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Hydroxide
CAS 1310-65-2
Molecular Formula HLiO
Quantity 25 g
Synonym Lithium Hydroxide, Lithium Hydrate
InChI Formula InChI=1S/Li.H2O/h;1H2/q+1;/p-1
SMILES [Li+].[OH-]
IUPAC Name lithium;hydroxide
Molecular Weight (g/mol) 23.95
Formula Weight 24.0187
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.