missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide, 99%, pure, anhydrous

Product Code. 10606422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606422 50 g Glass bottle 50g
10771861 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606422 Supplier Thermo Scientific Acros Supplier No. 203590500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-51-2 | ILi | 133.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium;iodide
SMILES [Li+].[I-]

Specifications

Boiling Point 1171.0°C
Melting Point 446.0°C
Physical Form Powder, Crystals and/or Granules
Quantity 50 g
Assay Percent Range 98.5% min. (on anhydrous substance) (Argentometry)
Linear Formula LiI
Fieser 01,615; 04,304; 05,410; 07,208; 09,283; 10,245; 11,300; 12,282; 15,185; 16,197; 17,85
Merck Index 15, 5591
Solubility Information Solubility in water: 165 g/100 mL (20°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Formula Weight 133.85
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Density 3.49
Chemical Name or Material Lithium iodide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium iodide, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : L-Limonene, 1, 8-Cineol, p-Cymene, D-Limonene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.