Learn More
Lithium iodide, anhydrous, 99.95% (metals basis)
Description
Used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor , which is a complex of Iodine is used in neuron detection , in an electrolyte of the dye-sensitized solar cell. It is an electrolyte additive for lithium-sulfur batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 1170°C to 1190°C |
| Melting Point | 449°C |
| Color | White |
| Physical Form | Crystalline, Powder |
| Quantity | 100 g |
| Linear Formula | LiI |
| Merck Index | 14,5535 |
| Solubility Information | Soluble in water,ethanol and acetone. |
| Formula Weight | 133.84 |
| Percent Purity | 99.95% |
| Show More |
Sicherheit und Handhabung
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.