missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide hydrate, 99%, extra pure

Product Code. 10441251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441251 100 g Glass bottle 100g
10286973 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441251 Supplier Thermo Scientific Acros Supplier No. 208011000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85017-80-7 | ILi | 133.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-80-7
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00149769
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci
PubChem CID 23688290
SMILES [Li+].[I-]

Specifications

Boiling Point 1171°C
Melting Point 73°C
Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Linear Formula LiI·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,freely soluble in acetone
IUPAC Name lithium;iodide;hydrate
Formula Weight 133.85
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Lithium iodide hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium iodide hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.