missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide hydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11386817
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11386817

Brand: Thermo Scientific Alfa Aesar 010738.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85017-80-7 | ILi | 133.84 g/mol

Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-80-7
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00149769
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci
PubChem CID 23688290
IUPAC Name lithium;iodide;hydrate
SMILES [Li+].[I-]

Specifications

Melting Point 73°C (in -H2O)
Physical Form Crystalline Aggregates
Quantity 25 g
Merck Index 14,5535
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 133.84 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Sensitivity Air and light sensitive; Hygroscopic
Assay (metals basis)
Chemical Name or Material Lithium iodide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.