missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide, ultra dry, 99.999% (metals basis)

Product Code. 11336878
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336878 5 g 5g
11346878 25 g 25g
11326878 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336878 Supplier Thermo Scientific Alfa Aesar Supplier No. 013600.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-51-2 | ILi | 133.84 g/mol

It is used as an absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium;iodide
SMILES [Li+].[I-]

Specifications

Boiling Point 1170°C to 1190°C
Melting Point 449°C
Color White
Physical Form Beads
Quantity 5 g
Merck Index 14,5535
Solubility Information Soluble in water,alcohol,acetone,methanol and ammonium hydoxide.
Formula Weight 133.84
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Refractive Index 1.955
Sensitivity Hygroscopic; Light sensitive
Density 4.076 g/mL
Assay (metals basis)
Chemical Name or Material Lithium iodide, Ultra dry
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.