missing translation for 'onlineSavingsMsg'
Learn More

Lithium isobutyrate, 98+%

Product Code. 11368138
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368138 500 g 500g
11348138 25 g 25g
11358138 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368138 Supplier Thermo Scientific Alfa Aesar Supplier No. 039372.36
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25179-23-1 | C4H7LiO2 | 94.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25179-23-1
Molecular Formula C4H7LiO2
Molecular Weight (g/mol) 94.038
MDL Number MFCD00210622
InChI Key IIDVGIFOWJJSIJ-UHFFFAOYSA-M
Synonym lithium isobutyrate, lithium 2-methylpropanoate, acmc-1ccd2, isobutyric acid lithium salt, lithium 1+ isobutyrate, lithium 1+ ion isobutyrate
PubChem CID 23665557
IUPAC Name lithium;2-methylpropanoate
SMILES [Li+].CC(C)C(=O)[O-]

Specifications

Quantity 500 g
Sensitivity Moisture sensitive
Formula Weight 94.03
Percent Purity ≥98%
Physical Form Powder or Lumps
Chemical Name or Material Lithium isobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.