Learn More
Lithium metaborate, anhydrous, 99.9% (metals basis)
Description
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 845°C |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 99.9% (metals basis) |
| Solubility Information | Soluble in ethanol. Partially soluble in cold water. |
| IUPAC Name | lithium;oxido(oxo)borane |
| Formula Weight | 49.75 |
| Sensitivity | Hygroscopic |
| Density | 1.397 g/mL |
| Chemical Name or Material | Lithium metaborate, anhydrous |
Safety and Handling
- Lithium metaborate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Lithium metaborate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.