missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol

Product Code. 11333528
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333528 100 mL 100mL
11323528 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333528 Supplier Thermo Scientific Alfa Aesar Supplier No. 039155.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 86745-52-0 | C12H30LiNbO6 | 370.212 g/mol

Lithium niobium ethoxide is used in the preparation of lithium niobate, which is a ferroelectric material used as an optical switch. It is also used as solid electrolytes due to its high ionic diffusivity in the field of material science.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86745-52-0
Molecular Formula C12H30LiNbO6
Molecular Weight (g/mol) 370.212
MDL Number MFCD00210625
InChI Key AVEKRHJBCCYCED-UHFFFAOYSA-N
Synonym lithium niobium ethoxide metals basis w/v in ethanol, lithium niobium ethoxide, lithium niobium 5+ ethoxide 1/1/6, niobium 5+ lithium 1+ hexakis ethoxide
PubChem CID 14693679
IUPAC Name lithium;ethanolate;niobium(5+)
SMILES [Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]

Specifications

Color Yellow
Flash Point 17°C (63°F)
Odor Alcohol-like
Quantity 100 mL
UN Number UN1170
Sensitivity Moisture sensitive
Formula Weight 370.21
Percent Purity ≥99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lithium niobium ethoxide
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.