missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
Description
Used as a precursor for chemical vapor deposition of thin films for lithium ion battery electrolyte materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Linear Formula | LiNb(OCH3)6 |
| Quantity | 100 mL |
| UN Number | UN1230 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Fully miscible with water. |
| Formula Weight | 285.92 |
| Percent Purity | ≥99% |
| Physical Form | Liquid |
| Assay | (metals basis) |
| Chemical Name or Material | Lithium niobium methoxide |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction