Learn More
Lithium nitrate hydrate, Puratronic™, 99.999% (metals basis)
Description
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 10 g |
| Assay Percent Range | 99.99% (metals basis) |
| Solubility Information | Soluble in water(2340g/L). Also soluble in ethanol,methanol,pyridine,ammonia and acetone. |
| IUPAC Name | lithium;nitrate |
| Formula Weight | 68.95 (Anhydrous) |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Lithium nitrate hydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.